Learn More
Cayman Chemical CaptoprIl DIsulfIde 10mg
SDP
Catalog No. 502940098
Encompass_Preferred
A metabolite and degradation product of captopril; a potential impurity in commercial preparations of captopril; inhibits angiotensin I-induced increases in mean arterial pressure in anesthetized dogs (ID50 = 0.231 mg/kg, i.v.)
Catalog No. 50-294-0098 Supplier Cayman Chemical Supplier No. 2801810 MG
A metabolite and degradation product of captopril; a potential impurity in commercial preparations of captopril; inhibits angiotensin I-induced increases in mean arterial pressure in anesthetized dogs (ID50 = 0.231 mg/kg, i.v.)