Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Captopril disulfide | 64806-05-9 | MFCD08063643 | 99.3% | 432.55 | C18H28N2O6S2 | 100MG
SDP

Catalog No. 5000235752
Encompass_Preferred

Captopril disulfide is the disulfide metabolite of the ACE inhibitor captopril with reported antihypertensive activity. It is supplied as an analytical standard and research reagent for analytical, pharmacological, and method development applications.

  • Analytical standard for research and analytical applications.
  • Disulfide metabolite of captopril with antihypertensive activity.
  • High purity (99.29%).
  • Molecular formula C18H28N2O6S2.
  • Molecular weight 432.55 g/mol.
  • Available in multiple sizes, including 100 mg.
  • Suitable for use as a reference material in method development and quality control.

Catalog No. 50-002-35752 Supplier Medchemexpress LLC Supplier No. HY123068100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Captopril disulfide is the disulfide metabolite of the ACE inhibitor captopril with reported antihypertensive activity. It is supplied as an analytical standard and research reagent for analytical, pharmacological, and method development applications.

  • Analytical standard for research and analytical applications.
  • Disulfide metabolite of captopril with antihypertensive activity.
  • High purity (99.29%).
  • Molecular formula C18H28N2O6S2.
  • Molecular weight 432.55 g/mol.
  • Available in multiple sizes, including 100 mg.
  • Suitable for use as a reference material in method development and quality control.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.