Learn More
Medchemexpress LLC Captopril disulfide | 64806-05-9 | MFCD08063643 | 99.3% | 432.55 | C18H28N2O6S2 | 5MG
SDP
Catalog No. 5000238700
Encompass_Preferred
Captopril disulfide is the symmetrical disulfide metabolite of the ACE inhibitor captopril, provided as a research-grade analytical standard for studies of angiotensin-converting enzyme inhibition and antihypertensive drug metabolism. Molecular formula C18H28N2O6S2, molecular weight 432.55, CAS 64806-05-9.
  • High reported purity (99.29%).
  • Analytical standard for metabolite and pharmacokinetic studies.
  • Applicable to ACE activity and antihypertensive research.
  • Available in milligram-scale pack sizes suitable for small-scale experiments.
  • Stable white solid with defined molecular identity.
Catalog No. 50-002-38700 Supplier Medchemexpress LLC Supplier No. HY1230685MG
Captopril disulfide is the symmetrical disulfide metabolite of the ACE inhibitor captopril, provided as a research-grade analytical standard for studies of angiotensin-converting enzyme inhibition and antihypertensive drug metabolism. Molecular formula C18H28N2O6S2, molecular weight 432.55, CAS 64806-05-9.
  • High reported purity (99.29%).
  • Analytical standard for metabolite and pharmacokinetic studies.
  • Applicable to ACE activity and antihypertensive research.
  • Available in milligram-scale pack sizes suitable for small-scale experiments.
  • Stable white solid with defined molecular identity.