Learn More
Medchemexpress LLC Captopril disulfide | 64806-05-9 | MFCD08063643 | 99.3% | 432.55 g/mol | C18H28N2O6S2 | 10MG

Supplier: Medchemexpress LLC HY12306810MG
Captopril disulfide is the disulfide dimer and metabolite of captopril with angiotensin-converting enzyme (ACE) inhibitory and antihypertensive activity. It is supplied as a research-grade reference material for biochemical and pharmacological studies of ACE pathways and cardiovascular function; supporting documentation is provided for quality and safety compliance.
- High analytical purity (99.3%).
- Molecular formula C18H28N2O6S2 and molecular weight 432.55 g/mol.
- Available in small research pack sizes (5 mg-100 mg), larger quantities available by quote.
- Documentation available: data sheet, certificate of analysis, and safety data sheet.
- Stored refrigerated at 4°C and protected from light.
- Intended for research use only; not for human or veterinary use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.