Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Captopril disulfide | 64806-05-9 | MFCD08063643 | 99.3% | 432.55 g/mol | C18H28N2O6S2 | 25MG
SDP

Catalog No. 5000238367
Encompass_Preferred

Captopril disulfide is a metabolite and disulfide dimer of captopril that exhibits antihypertensive activity. It is supplied as a research-grade analytical standard for laboratory use, suitable for biochemical assays, metabolic studies, and analytical reference work. Store solid material at 4°C protected from light; in solution, store at -80°C (up to 6 months) or -20°C (up to 1 month).

  • Research-grade analytical standard with documented antihypertensive activity.
  • Purity 99.3%.
  • Chemical formula C18H28N2O6S2.
  • Molecular weight 432.55 g/mol.
  • CAS number 64806-05-9.
  • Storage: solid at 4°C, protected from light; in solution at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-38367 Supplier Medchemexpress LLC Supplier No. HY12306825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Captopril disulfide is a metabolite and disulfide dimer of captopril that exhibits antihypertensive activity. It is supplied as a research-grade analytical standard for laboratory use, suitable for biochemical assays, metabolic studies, and analytical reference work. Store solid material at 4°C protected from light; in solution, store at -80°C (up to 6 months) or -20°C (up to 1 month).

  • Research-grade analytical standard with documented antihypertensive activity.
  • Purity 99.3%.
  • Chemical formula C18H28N2O6S2.
  • Molecular weight 432.55 g/mol.
  • CAS number 64806-05-9.
  • Storage: solid at 4°C, protected from light; in solution at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.