Learn More
Medchemexpress LLC Captopril EP impurity D (3-bromoisobutyric acid) | 56970-78-6 | 95.0% | 167.00 g·mol⁻¹ | C4H7BrO2 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01533210G
Encompass_Preferred
Captopril EP Impurity D (3-bromoisobutyric acid) is an analytical reference standard corresponding to an EP-listed impurity of the antihypertensive drug captopril. Supplied as a characterized solid, it is intended for impurity profiling, method development, and quality control of captopril formulations. Typical specifications include CAS 56970-78-6, molecular weight 167.00 g·mol⁻¹, and purity 95.0%.
  • EP-listed impurity standard for captopril
  • Intended for impurity profiling and method development
  • Suitable for quality control and analytical assays
  • Characterized solid with known molecular weight
  • Available in laboratory-scale package sizes
Catalog No. 50-003-02932