Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Captopril, MP Biomedicals

Catalog No. ICN15492801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15492801 1 g
ICN15492805 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15492801 Supplier MP Biomedicals, Inc Supplier No. 0215492801

Chemical Identifiers

CAS 62571-86-2
Molecular Formula C9H15NO3S
Molecular Weight (g/mol) 217.283
MDL Number MFCD00168073
InChI Key FAKRSMQSSFJEIM-RQJHMYQMSA-N
Synonym captopril, capoten, l-captopril, captopryl, lopirin, cesplon, captolane, tensoprel, acepress, captoril
PubChem CID 44093
ChEBI CHEBI:3380
IUPAC Name (2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid
SMILES CC(CS)C(=O)N1CCCC1C(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 217.3
Physical Form Powder
Chemical Name or Material Captopril
Recommended Storage Store at room temperature (15° to 30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.