Learn More
Sigma Aldrich Fine Chemicals Biosciences CaptoTM adhere Cytiva 17 5
Catalog No. 502904661
Encompass
A multimodal anion exchange resin designed for post-protein A purification of monoclonal antibodies at process scale. It can also be used in bind/elute mode.
Catalog No. 50-290-4661 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17544410
A multimodal anion exchange resin designed for post-protein A purification of monoclonal antibodies at process scale. It can also be used in bind/elute mode.