Learn More
Sigma Aldrich Fine Chemicals Biosciences CaptoTM Core 700 Cytiva 17
Catalog No. 502904074
Encompass
Novel core bead technology and multimodal octylamine ligand give Capto Core 700 dual functionality. This chromatography resin has both size exclusion and binding properties.
Catalog No. 50-290-4074 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17548101
Novel core bead technology and multimodal octylamine ligand give Capto Core 700 dual functionality. This chromatography resin has both size exclusion and binding properties.