Learn More
Sigma Aldrich Fine Chemicals Biosciences CaptoTM HiRes Q 5/50
Catalog No. 502908466
Encompass
Capto HiRes Q and Capto HiRes S are prepacked columns for high-reSOLUTIONon purification of proteins and other biomolecules by ion exchange chromatography. They are well-suited for the preparation of highly pure proteins for structural studies and for protein analysis.High reSOLUTIONon columns-for laboratory scale ion exchange chromatography.Suitable for preparing highly pure proteins-essential in for example structural studies using techniques such as cryo-electron microscopy or protein crystallography.Suitable for protein analysis in research or quality control-for example for characterization of target protein charge variants.High quality column-excellent reproducibility durability and high chemical and pH stabilities.Replacement for Mono Q and Mono S columns-delivering similar or improved reSOLUTIONon with easy transition from one column platform to the other.Capto HiRes ion exchange chromatography resinThe chromatography resins packed in Capto HiRes Q and
Catalog No. 50-290-8466 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE29275878
Capto HiRes Q and Capto HiRes S are prepacked columns for high-reSOLUTIONon purification of proteins and other biomolecules by ion exchange chromatography. They are well-suited for the preparation of highly pure proteins for structural studies and for protein analysis.High reSOLUTIONon columns-for laboratory scale ion exchange chromatography.Suitable for preparing highly pure proteins-essential in for example structural studies using techniques such as cryo-electron microscopy or protein crystallography.Suitable for protein analysis in research or quality control-for example for characterization of target protein charge variants.High quality column-excellent reproducibility durability and high chemical and pH stabilities.Replacement for Mono Q and Mono S columns-delivering similar or improved reSOLUTIONon with easy transition from one column platform to the other.Capto HiRes ion exchange chromatography resinThe chromatography resins packed in Capto HiRes Q and