Learn More
Sigma Aldrich Fine Chemicals Biosciences CaptoTM L Cytiva 17 5478 0
Catalog No. 502908685
Encompass
Capto(TM) L is a BioProcess chromatography medium (resin) for the capture of antibodies and antibody fragments. It combines a rigid high-flow agarose matrix with the immunoglobulin-binding recombinant protein L ligand which has a strong affinity to the variable region of antibody kappa light chains. Capto(TM) L is suitable for the capture of a wide range of antibody fragments such as Fabs single-chain variable fragments (scFv) and domain antibodies (Dabs).
Catalog No. 50-290-8685 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17547802
Add to Cart
Add to Cart
Request a Quote
Capto(TM) L is a BioProcess chromatography medium (resin) for the capture of antibodies and antibody fragments. It combines a rigid high-flow agarose matrix with the immunoglobulin-binding recombinant protein L ligand which has a strong affinity to the variable region of antibody kappa light chains. Capto(TM) L is suitable for the capture of a wide range of antibody fragments such as Fabs single-chain variable fragments (scFv) and domain antibodies (Dabs).