Learn More
Sigma Aldrich Fine Chemicals Biosciences CaptoTM Q Cytiva 17 5316 0
Catalog No. 502904213
Encompass
Capto(TM) Q is a strong anion exchange BioProcess medium designed to meet the demands of modern large-scale manufacturers for fast efficient and cost-effective capture and intermediate protein purification.Capto(TM) Q is designed for capture and intermediate purification of proteins from large feed volumes. It is composed of a rigid high-flow agarose matrix modified with dextran surface extenders and a strong quaternary ammonium (Q) anion exchanger. The combination of high volume throughput and high dynamic binding capacity for Capto(TM) Q raises the productivity in industrial downstream processes. Capto(TM) Q withstands effective and rigorous CIP procedure and the chemical stability ensures long media lifetime.Capto(TM) Q is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range Capto(TM) Q meets industrial demands with security of supply and comprehensive technical and
Catalog No. 50-290-4213 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17531602
Capto(TM) Q is a strong anion exchange BioProcess medium designed to meet the demands of modern large-scale manufacturers for fast efficient and cost-effective capture and intermediate protein purification.Capto(TM) Q is designed for capture and intermediate purification of proteins from large feed volumes. It is composed of a rigid high-flow agarose matrix modified with dextran surface extenders and a strong quaternary ammonium (Q) anion exchanger. The combination of high volume throughput and high dynamic binding capacity for Capto(TM) Q raises the productivity in industrial downstream processes. Capto(TM) Q withstands effective and rigorous CIP procedure and the chemical stability ensures long media lifetime.Capto(TM) Q is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range Capto(TM) Q meets industrial demands with security of supply and comprehensive technical and