Learn More
Sigma Aldrich Fine Chemicals Biosciences CaptoTM Q ImpRes Cytiva 17
Catalog No. 502904662
Encompass
Capto(TM) Q ImpRes is a strong anion exchange BioProcess medium designed to meet the demands of modern large-scale manufacturers for fast efficient and cost-effective intermediate and polishing protein purification.
Catalog No. 50-290-4662 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17547002
Capto(TM) Q ImpRes is a strong anion exchange BioProcess medium designed to meet the demands of modern large-scale manufacturers for fast efficient and cost-effective intermediate and polishing protein purification.