Learn More
Sigma Aldrich Fine Chemicals Biosciences CaptoTM SP ImpRes Cytiva 1
Catalog No. 502904218
Encompass
Capto(TM) SP ImpRes is a strong cation exchange BioProcess medium designed to meet the demands of modern large-scale manufacturers for fast efficient and cost-effective intermediate and polishing protein purification.
Catalog No. 50-290-4218 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17546802
Capto(TM) SP ImpRes is a strong cation exchange BioProcess medium designed to meet the demands of modern large-scale manufacturers for fast efficient and cost-effective intermediate and polishing protein purification.