Learn More
MOLECULAR DEPOT LLC Capture Buffer CLIA- 50 mL
Catalog No. 5000013530
Encompass_Preferred
Capture Antibody Stabilization Buffer consists of a chemiluminescence buffer extensively tested for the stabilization of a wide variety of capture proteins and antibodies This buffer contains a proprietary collection of proteins detergents and other chemicals that promote the long-term stability of magnetic beads For in vitro research use only
Catalog No. 50-000-13530 Supplier MOLECULAR DEPOT LLC Supplier No. B2010187
Capture Antibody Stabilization Buffer consists of a chemiluminescence buffer extensively tested for the stabilization of a wide variety of capture proteins and antibodies This buffer contains a proprietary collection of proteins detergents and other chemicals that promote the long-term stability of magnetic beads For in vitro research use only