Learn More
Cederlane CAPTUREome S-Palmitoylated Protein Kit, Unit Size 1 Kit, Assay Method Acyl-RAC, Sample Capacity up to 20 ~1 mg samples, Storage Temperature 4°C, Shelf Life 6 months
SDP
Supplier:  Cederlane K010311
Encompass
CAPTUREome S-Palmitoylated Protein Kit, Unit Size 1 Kit, Assay Method Acyl-RAC, Sample Capacity up to 20 ~1 mg samples, Storage Temperature 4°C, Shelf Life 6 months
Catalog No. NC2258288