Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ AAT-XL Ligand Leakage ELISA
GREENER_CHOICE

Catalog No. 810383210 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 assay x 96-well plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81-038-3210 10 assay x 96-well plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 81-038-3210 Supplier Thermo Scientific™ Supplier No. 810383210
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Thermo Scientific™ CaptureSelect™ AAT-XL Ligand Leakage ELISA is designed to detect possible leached ligand of alpha-1 anti-trypsin (AAT) purified using CaptureSelect AAT-XL Affinity Matrix.

CaptureSelect AAT-XL Ligand Leakage ELISA is designed to detect possible leached ligand of alpha-1 anti-trypsin (AAT) purified using CaptureSelect AAT-XL Affinity Matrix. The AAT-XL affinity ligand is a 13-kD recombinant single-domain antibody fragment (VHH affinity ligand) expressed in yeast and covalently coupled to a chromatography matrix for high chemical stability and low leakage. During the chromatography process small amounts of ligand can leach from the chromatography support and co-elute with the target protein.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand concentrations greater than or equal to 1 ng/mL. The CaptureSelect AAT-XL Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

Specifications

Content And Storage Store at -20°C
Detection Method Absorbance at 450 nm
Immunoassay Kit Format Sandwich ELISA Kit
Product Line CaptureSelect
Product Type Ligand Leakage ELISA
Sample Type Ligand Leachate
For Use With (Application) Ligand Leakage ELISA assay
Ligand Type CaptureSelect Human Alpha-1 Antitrypsin (AAT) Affinity Ligand
Quantity 10 assay x 96-well plate
Shipping Condition Dry Ice
Target Target-specific ELISA reagents
Detection Limit 1 ng/mL
Format Individual components
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.