Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ AAT-XL RoboColumn™ Pre-packed columns

Catalog No. 5943832200 Shop All Thermo Scientific Products
Change view
Click to view available options
Bed Volume:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Volume
59-438-32200 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 59-438-32200 Supplier Thermo Scientific™ Supplier No. 5943832200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ CaptureSelect™ RoboColumn™ are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

CaptureSelect RoboColumn™ are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

The CaptureSelect AAT-XL Affinity Matrix in this RoboColumn has been specifically designed for the purification of native human alpha-1 antitrypsin (AAT) from complex sources like plasma/serum and recombinant human AAT from cell culture supernatants. The high selectivity and yield obtained using CaptureSelect AAT-XL Affinity Matrix enable a robust and efficient purification process with excellent purity obtained in a single step.

Features of this affinity matrix include:

  • Highly selective for native and recombinant human AAT
  • High recovery and yield obtained in a single purification step
  • Mild elution at pH 6
  • Excellent scalability
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Characteristics

  • Column size: 200 μL, 1 strip, 8 columns (8-row)
  • Column dimension: 5-mm diameter, 10-mm bed height
  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect AAT-XL affinity ligand
  • Ligand coupling: epoxide coupling
  • Binding capacity: 15 mg AAT/mL resin depending on flow rate, column height, and contact time
  • Elution conditions: 20 mM Tris, 2.0 M MgCl2, pH 6.0
  • Formulation buffer: 20% (v/v) ethanol
  • Operating pressure: 2 bar (maximum)
  • Flow rate: 16–150 cm/hr

RoboColumn is a trademark of Repligen.

Specifications

Max. Pressure 2 bar
Pore Size Not Applicable
Column Capacity 200 μL
Length (Metric) 1 cm
Quantity 1 Strip, 8 Columns
Particle Size 65 μm
For Use With (Equipment) Chromatography Systems
Type Affinity Column
Diameter (Metric) Inner 0.5 cm
Content And Storage Store at 2°C to 8°C. Protect from light. Do not freeze.
Product Line CaptureSelect
Ligand Type CaptureSelect Human Alpha-1 Antitrypsin (AAT) Affinity Ligand
Matrix Agarose
Shipping Condition Wet Ice
Column Format Pre-packed RoboColumn
Bed Volume 200 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.