Learn More
Thermo Scientific™ CaptureSelect™ AAVX Ligand Leakage ELISA

Description
CaptureSelect AAVX Ligand Leakage ELISA is designed to detect possible leached ligand of adeno-associated viruses (AAV) purified using POROS CaptureSelect AAVX Affinity Matrix. The ligand, a 13-kD recombinant protein, is covalently coupled for high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the purified AAV.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. POROS CaptureSelect AAVX Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Specifications
Specifications
| Content And Storage | 3 x 1 mL, store at -5°C to -30°C. |
| Product Line | CaptureSelect |
| Product Type | ELISA Kit |
| Quantity | 10 Assays |
| Shipping Condition | Dry Ice |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.