Learn More
Thermo Scientific™ CaptureSelect™ Alpha-1 Antitrypsin Select Leakage ELISA

Description
CaptureSelect Alpha-1 AntitrypsinSelect Leakage ELISA is designed to detect possible leached ligand of human Alpha-1 antitrypsin (AAT) purified using AAT Select affinity resin (Cytiva). Even when covalently attached, small amounts of the Alpha-1 Antitrypsin Select affinity ligand can leach off of the chromatography support and co-elute with the purified product.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect Alpha-1 Antitrypsin Select Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Specifications
Specifications
| Content And Storage | Store at -5°C to -30°C. |
| Assay | ELISA |
| For Use With (Equipment) | Microplate Reader |
| Product Line | CaptureSelect |
| Quantity | 1 Assay |
| Target | Alpha-1 AntitrypsinSelect Affinity Ligand |
| Type | ELISA Kit |
| Format | Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.