Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ Alpha-1 Antitrypsin Select Leakage ELISA
GREENER_CHOICE

Catalog No. 810287010 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Assay
10 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81-028-7010 10 Assays
81-028-7001 1 Assay
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 81-028-7010 Supplier Thermo Scientific™ Supplier No. 810287010
Add to Cart
Add to Cart
Request a Quote

Thermo Scientific™ CaptureSelect™ Alpha-1 AntitrypsinSelect Leakage ELISA is designed to detect possible leached ligand of human Alpha-1 antitrypsin (AAT) purified using AAT Select affinity resin (Cytiva).

CaptureSelect Alpha-1 AntitrypsinSelect Leakage ELISA is designed to detect possible leached ligand of human Alpha-1 antitrypsin (AAT) purified using AAT Select affinity resin (Cytiva). Even when covalently attached, small amounts of the Alpha-1 Antitrypsin Select affinity ligand can leach off of the chromatography support and co-elute with the purified product.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect Alpha-1 Antitrypsin Select Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

Specifications

Content And Storage Store at -20°C.
Assay ELISA
Detection Method Absorbance at 450 nm
Immunoassay Kit Format Sandwich ELISA Kit
Product Line CaptureSelect
Product Type Ligand Leakage ELISA
Sample Type Ligand Leachate
Target Species Not Applicable
For Use With (Equipment) Microplate Reader
Quantity 10 Assays
Shipping Condition Dry Ice
Target Alpha-1 AntitrypsinSelect Affinity Ligand
Detection Limit 1 ng/mL
Format Individual components
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.