Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ Antithrombin III Affinity Matrix
GREENER_CHOICE

Catalog No. 190317005
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-031-7005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-031-7005 Supplier Thermo Scientific™ Supplier No. 190317005
Add to Cart
Add to Cart

This is one of a group of Thermo Scientific™ CaptureSelect™ protein purification affinity resins developed for the simple, single-step purification of a variety human plasma and serum proteins.

This is one of a group of CaptureSelect protein purification affinity resins developed for the simple, single-step purification of a variety human plasma and serum proteins. This unique selection of affinity purification resins can purify proteins or other non-antibody biomolecules from either recombinant or native sources with a high degree of affinity and specificity, and are immobilized on a high capacity agarose support.

Specifications

Content And Storage Store at 2°C to 8°C.
Form Liquid Suspension
Product Line CaptureSelect
Quantity 5 mL
Type Bulk Resin
Stationary Phase Anti Antithrombin III
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.