Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-AAVX Conjugate
GREENER_CHOICE

Catalog No. 7103522500
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-035-22500 500 μg
71-035-22100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-035-22500 Supplier Thermo Scientific™ Supplier No. 7103522500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ CaptureSelect™ Biotin Anti-AAVX Conjugate consists of a 14 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to native and recombinant adeno-associated virus (AAV) particles of various serotypes.

CaptureSelect Biotin Anti-AAVX Conjugate consists of a 14 kDa recombinant single domain antibody fragment that specifically binds with high affinity and selectivity to native and recombinant adeno-associated virus (AAV) particles of various serotypes.

The anti-AAVX affinity ligand has demonstrated binding reactivity towards a large set of different AAV serotypes including AAV1 to AAV8, and AAVrh10, indicating that an even broader range of AAV serotypes could potentially be recognized and detected. The affinity ligand is chemically conjugated to biotin via a spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

This conjugate enables the detection, quantitation, and characterization of native and recombinant AAV virus particles. Applications for CaptureSelect Biotin Anti-AAVX Conjugate include ELISA, immunoprecipitation with streptavidin-coupled Dynabeads, Gyrolab™-based immunoassays, and use with label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Content And Storage Store at -5°C to -30°C.
Shipping Condition Dry Ice
Form Liquid
Product Line CaptureSelect
Quantity 500 μg
Type Conjugate
Ligand Type Antibody
How many biotin molecules are there per antibody molecule in your CaptureSelect Biotin Anti-AAVX Conjugate (Cat. No. 7103522500)?

All our CaptureSelect biotin conjugates are directionally conjugated with a maximal degree of labeling of 2 biotin molecules per VHH.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.