Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-FVIII Conjugate
GREENER_CHOICE

Catalog No. 7102862500
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-028-62500 500 μg
71-028-62100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-028-62500 Supplier Thermo Scientific™ Supplier No. 7102862500
Get an Extra Analytical Consumable with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ CaptureSelect™ Biotin Anti-FVIII Conjugate consists of a 13 kDa recombinant single domain antibody fragment with high affinity and selectivity for both recombinant (full length and B-domain deleted) and plasma derived human Factor VIII (FVIII), through recognition of the human FVIII light chain.

CaptureSelect Biotin Anti-FVIII Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for both recombinant (full length and B-domain deleted) and plasma derived human Factor VIII (FVIII), through recognition of the human FVIII light chain. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect Biotin Anti-FVIII Conjugate allows you to:

  • Detect, quantitate, and characterize recombinant and plasma derived human FVIII

Applications for CaptureSelect Biotin Anti-FVIII Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Product Line CaptureSelect
Quantity 500 μg
Type Conjugate
Label or Dye Biotin & Other Haptens
Ligand Type Antibody

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.