Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ Bovine IgA Affinity Matrix
GREENER_CHOICE

Catalog No. 2943511010
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-435-11010 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 29-435-11010 Supplier Thermo Scientific™ Supplier No. 2943511010
Add to Cart
Add to Cart

Thermo Scientific™ CaptureSelect™ Bovine IgA Affinity Matrix has been designed specifically for the purification of bovine IgA from whey.

CaptureSelect Bovine IgA Affinity Matrix has been designed specifically for the purification of bovine IgA from whey.

Features of this affinity matrix include:

  • Binds monomeric, dimeric, and secretory bovine IgA
  • One-step purification from whey
  • Excellent scalability

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, aldehyde-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect Bovine IgA affinity ligand
  • Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
  • Binding capacity: ∼10 g/L resin
  • Elution conditions: 0.1 M citric acid pH 3.0 or 0.1M glycine pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Binding Capacity ∼10 g/L resin
Flow Rate 50 to 200 cm/hr. (Up to 2 bar)
Matrix Agarose-based, aldehyde-activated
Content And Storage Store at 2°C to 8°C.
Particle Size 65 ±10 μm
Form Liquid Suspension
Product Line CaptureSelect
Quantity 10 mL
Type Affinity Matrix
Ligand Type Bovine IgA affinity ligand
Stationary Phase Anti Bovine IgA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.