Learn More
Description
CaptureSelect C1-InhXL Ligand Leakage ELISA is designed to detect possible leached ligand of C1-esterase inhibitor (C1Inh) purified using POROS CaptureSelect C1-InhXL Affinity Resin. The C1-InhXL affinity ligand is a 12–15 kD recombinant single-domain antibody fragment (VHH affinity ligand) expressed in yeast and covalently coupled to a chromatography matrix for high chemical stability and low leakage. During the chromatography process small amounts of the ligand can leach from the chromatography support and co-elute with the target protein.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect C1-InhXL Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Specifications
Specifications
| Content And Storage | Store in freezer (-5°C to -30°C). |
| Detection Method | Absorbance at 450 nm |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Line | CaptureSelect |
| Product Type | Ligand Leakage ELISA |
| Sample Type | Ligand Leachate |
| Ligand Type | CaptureSelect C1-InhXL Affinity Ligand |
| Quantity | 10 Assays |
| Shipping Condition | Dry Ice |
| Target | CaptureSelect C1-InhXL Affinity Ligand |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
