Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ CH1-XL Ligand Leakage ELISA
GREENER_CHOICE

Catalog No. 810346201
Change view
Click to view available options
Quantity:
1 Assay
10 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81-034-6201 1 Assay
81-034-6210 10 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 81-034-6201 Supplier Thermo Scientific™ Supplier No. 810346201
Only null left
Add to Cart
Add to Cart

Thermo Scientific™ CaptureSelect™ CH1-XL Ligand Leakage ELISA Kit is designed to detect possible leached ligand of antibodies or antibody fragments purified using CaptureSelect CH1-XL Affinity Matrix.

CaptureSelect CH1-XL Ligand Leakage ELISA Kit is designed to detect possible leached ligand of antibodies or antibody fragments purified using CaptureSelect CH1-XL Affinity Matrix. The ligand, a 14-kD recombinant protein, is covalently coupled for high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect CH1-XL Ligand Leakage ELISA Kit can be used as a tool to aid in development of an optimal purification process and in routine quality control of in-process streams and final product.

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Shipping Condition Dry Ice
Product Line CaptureSelect
Quantity 1 Assay
Type ELISA Kit
Format Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.