Learn More
Description
CaptureSelect CH1-XL Ligand Leakage ELISA Kit is designed to detect possible leached ligand of antibodies or antibody fragments purified using CaptureSelect CH1-XL Affinity Matrix. The ligand, a 14-kD recombinant protein, is covalently coupled for high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect CH1-XL Ligand Leakage ELISA Kit can be used as a tool to aid in development of an optimal purification process and in routine quality control of in-process streams and final product.
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| Detection Method | Absorbance at 450 nm |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Line | CaptureSelect |
| Product Type | Ligand Leakage ELISA |
| Sample Type | Ligand Leachate |
| Target Species | Not Applicable |
| Quantity | 10 Assays |
| Shipping Condition | Dry Ice |
| Target | CaptureSelect CH1-XL Affinity Ligand |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
