Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ CH1-XL Pre-packed Column

Catalog No. 494346205 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 x 5 mL Column
5 x 1 mL Columns
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-434-6205 1 x 5 mL Column
49-434-6201 5 x 1 mL Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 49-434-6205 Supplier Thermo Scientific™ Supplier No. 494346205

missing translation for 'validMainframeMsg'

Thermo Scientific™ CaptureSelect™ CH1-XL Affinity Matrix is an improved version of CaptureSelect IgG-CH1 Affinity Matrix, featuring a wider elution range (pH 3 to 4.5), increased binding capacity, and improved stability at higher pH, offered here in pre-packed format.

CaptureSelect CH1-XL Affinity Matrix is an improved version of CaptureSelect IgG-CH1 Affinity Matrix, featuring a wider elution range (pH 3.0–4.5), increased binding capacity, and improved stability at higher pH, offered here in pre-packed format.

CaptureSelect CH1-XL Affinity Matrix recognizes the CH1 domain of human IgG antibodies, enabling purification of all subclasses (1, 2, 3, and 4) of human IgG and Fab fragments, independent of the light-chain iso-type and independent of the source material. Due to its unique selectivity for the CH1 domain, no co-purification of free light-chain contaminants will occur. Since no cross-binding is observed for bovine IgG, the product can be used for the purification of IgGs and antibody fragments in the presence of fetal calf serum.

The affinity matrix is offered in a pre-packed format and consists of 5 x 1 mL, or 1 x 5 mL columns with connectors, that are compatible with standard chromatography equipment.

Specifications

Content And Storage Store in refrigerator (2°C to 8°C).
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Product Line CaptureSelect
Quantity 1 x 5 mL Column
Type Pre-packed Column
Ligand Type CH1-XL ligand
Stationary Phase CH1-XL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.