Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ CH1-XL RoboColumn™ Pre-packed Column
GREENER_CHOICE

Catalog No. 5943462200
Catalog No. 59-434-62200 Supplier Thermo Scientific™ Supplier No. 5943462200
Add to Cart
Add to Cart

Thermo Scientific™ CaptureSelect™ RoboColumn™ small chromatography columns are used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

CaptureSelect RoboColumn™ small chromatography columns are used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

CaptureSelect CH1-XL Affinity Matrix recognizes the CH1 domain of human IgG antibodies, thereby enabling purification of all Fab fragments, independent of the light-chain type (kappa or lambda) and without binding of over-expressed light chain and light-chain dimers. In addition, it can be used for the purification of human IgGs from all four subclasses (1, 2, 3, and 4).

Features of this affinity matrix include:

  • Platform approach for purification of high quality and correctly assembled human Fab fragments
  • No co-purification of over-expressed free light chains due to selectivity for the CH1 domain
  • Elution possible at pH 4.0–4.5 with 50 mM acetate buffer
  • Caustic cleaning with up to 50 mM NaOH
  • Excellent scalability
  • No cross-binding to bovine IgG, therefore presence of fetal bovine serum is not a problem
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Characteristics

  • Column size: 200 μL and 600 μL, 1 strip, 8 columns (8-row)
  • Column dimension: 200 μL: 5 x 10 mm
  • Column dimension: 600 μL: 5 x 30 mm
  • Matrix: agarose based, epoxide-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect CH1-XL affinity ligand
  • Ligand coupling: epoxide coupling
  • Binding capacity: >15 mg/mL human Fab fragment depending on flow rate, column height, contact time. For humanized Fabs, lower capacities may be observed.
  • Elution conditions: 100 mM glycine buffer, pH 3.0; 50 mM acetate buffer, pH 4.0–4.5
  • Formulation buffer: 20% (v/v) ethanol
  • Operating pressure: 2 bar (maximum)
  • Flow rate: 16–150 cm/h

RoboColumn is a trademark of Repligen.

Specifications

Flow Rate 16 to 150 cm/hr.
Max. Pressure 2 bar
Stationary Phase CH1-XL affinity ligand on agarose bead
Column Capacity 200 μL
Length (Metric) 10 mm
Quantity 200 μL
Particle Size 65 ±10 μm
For Use With (Application) Chromatography
Type RoboColumn
Diameter (Metric) Inner 5 mm
Content And Storage Store in refrigerator (2°C to 8°C)
Binding Capacity >15 mg/mL human Fab fragment
Product Line CaptureSelect
Ligand Type CH1-XL affinity ligand
Matrix Agarose-based, Epoxide-activated
Shipping Condition Room temperature, or on wet ice
Column Format 8-well Strips
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.