Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ FcXP RoboColumn™ Pre-packed columns

Catalog No. 5943712200 Shop All Thermo Scientific Products
Change view
Click to view available options
Bed Volume:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Volume
59-437-12200 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 59-437-12200 Supplier Thermo Scientific™ Supplier No. 5943712200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ CaptureSelect™ RoboColumn™ are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

CaptureSelect RoboColumn™ are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

CaptureSelect FcXP affinity matrix recognizes the CH3 domain of all human IgG subclasses (IgG1, 2, 3, and 4), enabling purification of all IgG subclasses and Fc fusion proteins directly from complex source materials in a robust and efficient purification process with excellent purity obtained in a single step.

Features of tFcXP affinity matrix include:

  • Mild elution, making it suitable for Fc fusion proteins
  • One-step purification from crude material with high purity and yield
  • Human specific, no binding to bovine antibodies
  • Excellent scalability
  • Non–animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Characteristics

  • Column size: 200 μL, 1 strip, 8 columns (8-row)
  • Column dimension: 5-mm diameter, 10-mm bed height
  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect FcXP affinity ligand
  • Ligand coupling: epoxide coupling
  • Binding capacity: 40-50 g IgG/L resin depending on flow rate, column height, and contact time
  • Elution conditions: 20 mM citric acid or acetic acid, pH 3–4
  • Formulation buffer: 20% (v/v) ethanol
  • Operating pressure: 2 bar (maximum)
  • Flow rate: 16–150 cm/hr

Specifications

Flow Rate 16 to 150 cm/hr.
Max. Pressure 2 bar
Pore Size Not Applicable
Column Capacity 200 μL
Length (Metric) 1 cm
Quantity 1 Strip, 8 Columns
Particle Size 65 μm
For Use With (Equipment) Chromatography Systems
Type Affinity Column
Diameter (Metric) Inner 0.5 cm
Content And Storage Store at 2°C to 8°C. Protect from light. Do not freeze.
Binding Capacity 40 to 50 g IgG/L Resin
Product Line CaptureSelect
Ligand Type CaptureSelect FcXP Affinity Ligand
Matrix Agarose
Shipping Condition Wet Ice
Column Format Pre-packed RoboColumn
Bed Volume 200 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.