Learn More
Thermo Scientific™ CaptureSelect™ FSH Affinity Matrix

Description
CaptureSelect FSH Affinity Matrix has been specifically designed for the purification of follicle stimulating hormone (FSH) from recombinant sources.
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Specifications
Specifications
| Flow Rate | 200 to 400 cm/hr. |
| Matrix | Agarose-based, aldehyde-activated |
| Content And Storage | Store at 2°C to 8°C. |
| Particle Size | 65 μm |
| Form | Liquid Suspension |
| Product Line | CaptureSelect |
| Quantity | 50 mL |
| Type | Affinity Matrix |
| Stationary Phase | Anti-Follicle Stimulating Hormone |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.