Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ GM-CSF Affinity Matrix
GREENER_CHOICE

Catalog No. p-7180774
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-434-72005 5 mL
29-434-72010 10 mL
29-434-72050 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 29-434-72005 Supplier Thermo Scientific™ Supplier No. 2943472005
Only null left
Add to Cart
Add to Cart

Thermo Scientific™ CaptureSelect™ GM-CSF Affinity Matrix is suitable for the purification of granulocyte-macrophage colony-stimulating factor (GM-CSF), a cytokine that functions as a white blood cell growth factor.

CaptureSelect GM-CSF Affinity Matrix is suitable for the purification of granulocyte-macrophage colony-stimulating factor (GM-CSF), a cytokine that functions as a white blood cell growth factor. The affinity matrix enables purification of this cytokine with high purity and yield in a single step. The CaptureSelect affinity ligand is immobilized on a high-capacity agarose matrix.

Specifications

Content And Storage Store in refrigerator (2°C to 8°C).
Shipping Condition Room Temperature
Form Liquid Suspension
Product Line CaptureSelect
Quantity 5 mL
Type Affinity Matrix
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.