Learn More
Thermo Scientific™ CaptureSelect™ hCG Affinity Matrix

Description
CaptureSelect hCG Affinity Matrix was specifically designed for the purification of human chorionic gonadotropin (hCG) from recombinant sources.
Some features of the CaptureSelect hCG Affinity Matrix include:
- Selective for hCG, with cross-binding to LH, FSH, and TSH
- Enables efficient purification of recombinant gonadotropins with high yields and purity
- Offers excellent scalability
- Non–animal-derived
Human gonadotropins (hCG, LH, FSH, TSH) are glycoprotein hormones secreted by gonadotrope cells of the pituitary gland that are often produced as recombinant proteins. They have an identical alpha chain subunit that is recognized by the ligand. Consequently, the CaptureSelect hCG Affinity Matrix shows binding to all human gonadotropins. The high selectivity and yields obtained using CaptureSelect hCG Affinity Matrix enable a robust and efficient purification process for hCG with excellent purity obtained in one step.
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Main characteristics
- Matrix: agarose-based, aldehyde-activated
- Average particle size: 65 ± 10 μm
- Ligand: CaptureSelect hCG affinity ligand
- Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
- Binding capacity: approx. 4 mg/mL resin (determined with HCG)
- Elution conditions: 20 mM citric acid, pH 3.0
- Flow characteristics: 50–200 cm/h (up to 2 bar)
- Formulation buffer: 20% (v/v) ethanol
Specifications
Specifications
| Content And Storage | Store in refrigerator (2°C to 8°C). |
| Shipping Condition | Room Temperature |
| Stationary Phase | Anti-gonadotropin |
| Product Line | CaptureSelect |
| Type | Affinity Matrix |
| Quantity | 5 mL |
| Form | Agarose Bead |
| Particle Size | 65 ± 10 μm, 65 ±10 μm |
| Binding Capacity | 4 mg/mL |
| Flow Rate | 50 to 200 cm/hr. (Up to 2 bar) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.