Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ hCG Affinity Matrix
GREENER_CHOICE

Catalog No. 194341050
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
250 mL
1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-434-1050 50 mL
19-434-1005 5 mL
19-434-1010 10 mL
19-434-10250 250 mL
19-434-101L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-434-1050 Supplier Thermo Scientific™ Supplier No. 194341050
Only null left
Add to Cart
Add to Cart
Request a Quote

Thermo Scientific™ CaptureSelect™ hCG Affinity Matrix was specifically designed for the purification of human chorionic gonadotropin (hCG) from recombinant sources.

CaptureSelect hCG Affinity Matrix was specifically designed for the purification of human chorionic gonadotropin (hCG) from recombinant sources.

Some features of the CaptureSelect hCG Affinity Matrix include:

  • Selective for hCG, with cross-binding to LH, FSH, and TSH
  • Enables efficient purification of recombinant gonadotropins with high yields and purity
  • Offers excellent scalability
  • Non–animal-derived

Human gonadotropins (hCG, LH, FSH, TSH) are glycoprotein hormones secreted by gonadotrope cells of the pituitary gland that are often produced as recombinant proteins. They have an identical alpha chain subunit that is recognized by the ligand. Consequently, the CaptureSelect hCG Affinity Matrix shows binding to all human gonadotropins. The high selectivity and yields obtained using CaptureSelect hCG Affinity Matrix enable a robust and efficient purification process for hCG with excellent purity obtained in one step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Matrix: agarose-based, aldehyde-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect hCG affinity ligand
  • Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
  • Binding capacity: approx. 4 mg/mL resin (determined with HCG)
  • Elution conditions: 20 mM citric acid, pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Content And Storage Store in refrigerator (2°C to 8°C).
Shipping Condition Room Temperature
Stationary Phase Anti-gonadotropin
Product Line CaptureSelect
Type Affinity Matrix
Quantity 50 mL
Form Agarose Bead
Particle Size 65 ± 10 μm, 65 ±10 μm
Binding Capacity 4 mg/mL
Flow Rate 50 to 200 cm/hr. (Up to 2 bar)
Ligand Type HCG affinity ligand
Matrix Agarose-based, aldehyde-activated
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.