Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ HRP Anti-AAV9 Conjugate
GREENER_CHOICE

Catalog No. 7303332500
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-033-32500 500 μL
73-033-32100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 73-033-32500 Supplier Thermo Scientific™ Supplier No. 7303332500
Get an Extra Analytical Consumable with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Thermo Scientific™ CaptureSelect™ HRP Anti-AAV9 Conjugate includes a 13-kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds adeno-associated virus particles of serotype 9 (AAV9).

CaptureSelect HRP Anti-AAV9 Conjugate includes a 13-kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds adeno-associated virus particles of serotype 9 (AAV9).

CaptureSelect HRP Anti-AAV9 Conjugate can be combined with CaptureSelect Biotin Anti-AAV9 Conjugate (Cat. No. 7103332100) in enzyme-linked immunoassays to quantify the total number of AAV9 capsids. Sandwich ELISA set-ups can be designed using the biotin anti-AAV9 conjugate as a capture reagent immobilized on a streptavidin coated surface. HRP anti-AAV9 conjugate is then used for detection of bound AAV9 capsids.

For assay development to measure total capsids for other AAV serotypes, we recommend CaptureSelect Biotin Anti-AAVX Conjugate (Cat. No. 7103522100) and CaptureSelect HRP Anti-AAVX Conjugate (Cat. No. 7303522100).

Product details

  • Format type: HRP, liquid
  • Buffer: PBS, 50% (v/v) glycerol
  • Storage: in freezer (-5 to -30°C)
  • Concentration: 0.5 mg/mL
  • Volume: 0.1 mL
  • Recommended working dilution for sandwich ELISA: 1:5000 to 1:10,000 (the optimal working dilution should be determined empirically for each application)

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Concentration 0.5 mg/mL
Content And Storage Store in freezer (-5°C to -30°C).
Form Liquid
Product Line CaptureSelect
Quantity 500 μL
For Use With (Application) Quantitation of AAV9 capsids
Type Conjugate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.