Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ Human Albumin Affinity Matrix
GREENER_CHOICE

Catalog No. 191297050 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-129-7050 50 mL
19-129-7005 5 mL
19-129-7010 10 mL
19-129-701L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-129-7050 Supplier Thermo Scientific™ Supplier No. 191297050
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Thermo Scientific™ CaptureSelect™ Human Albumin Affinity Matrix has been specifically designed for the purification of human albumin and albumin fusion proteins.

CaptureSelect Human Albumin Affinity Matrix has been specifically designed for the purification of human albumin and albumin fusion proteins.

Features of this affinity matrix include:

  • Highly selective for human albumin
  • Unique elution characteristics allowing for gentle elution of albumin fusion proteins
  • Excellent scalability
  • Non-animal-derived

The high selectivity and yields obtained using CaptureSelect Human Albumin Affinity Matrix enable a robust and efficient purification process with excellent purity obtained in one step. The mild elution conditions make the resin suitable for the purification of albumin fusion proteins, especially those that are pH sensitive.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main Characteristics:

  • Matrix: agarose-based, aldehyde-activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect HSA affinity ligand
  • Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
  • Binding capacity: ∼15 mg albumin/mL resin
  • Elution conditions (neutral pH): 20 mM Tris, pH 7.4 with 2 M MgCl2 or with 1 M NaCl + 50%(v/v) propylene glycol or with 1 M NaCl + 0.5 M L-arginine
  • Elution conditions (acidic pH): 0.1 M glycine, pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20%(v/v) ethanol

Specifications

Binding Capacity ∼15 mg albumin/mL resin
Capacity (Metric) ∼15 mg albumin/mL resin
Flow Rate 50 to 200 cm/hr. (Up to 2 bar)
Matrix Agarose-based, aldehyde-activated
Content And Storage • Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light
Particle Size 65 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 50 mL
Type Affinity Matrix
Ligand Type HSA affinity ligand
Stationary Phase Anti-HSA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.