Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ IgG-Fc (Multispecies) Affinity Matrix
GREENER_CHOICE

Catalog No. 2942852050 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-428-52050 50 mL
29-428-52005 5 mL
29-428-52010 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 29-428-52050 Supplier Thermo Scientific™ Supplier No. 2942852050
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Thermo Scientific™ CaptureSelect™ IgG-Fc (Multispecies) Affinity Matrix has been specifically designed to purify IgG through high-affinity binding of the IgG Fc region.

CaptureSelect IgG-Fc (Multispecies) Affinity Matrix has been specifically designed to purify IgG through high-affinity binding of the IgG Fc region. This resin can be used for purification of IgG from a broad range of species. The tested species and their specificity are shown in the figure below.

Features of this affinity matrix include:

  • One-step IgG purification with high purity, from crude material
  • Excellent scalability
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect IgG-Fc (multispecies) affinity ligand
  • Ligand coupling method: epoxide coupling
  • Binding capacity: >25 g IgG/L resin tested with human IgG
  • Elution conditions: 0.1 M glycine, pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Flow Rate 50 to 200 cm/hr. (Up to 2 bar)
Matrix Agarose
Content And Storage Store at 2°C to 8°C in a light-protected environment. Avoid direct exposure to sunlight. Do not freeze
Max. Operating Pressure 2 bar
Shipping Condition Wet Ice
Particle Size 65 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 50 mL
Type Affinity Matrix
Ligand Type CaptureSelect IgG-Fc (multispecies) Affinity Ligand
Binding Capacity >25 g IgG/L Resin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.