Learn More
Thermo Scientific™ CaptureSelect™ IgG3 (Hu) Affinity Matrix

Description
CaptureSelect IgG3 (Hu) Affinity Matrix can be used for the purification and isolation of IgG3, without cross-reactivity to other immunoglobulins, like other human IgG subclasses, IgA, and IgM, from complex sources like plasma, serum, and cell culture supernatants.
Specifications
Specifications
| Content And Storage | Store at 2°C to 8°C. |
| Form | Liquid Suspension |
| Product Line | CaptureSelect |
| Quantity | 50 mL |
| Type | Affinity Matrix |
| Stationary Phase | Anti-IgG3 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.