Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ IgG3 (Hu) Affinity Matrix
GREENER_CHOICE

Catalog No. A191304005 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A191304005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A191304005 Supplier Thermo Scientific™ Supplier No. 191304005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ CaptureSelect™ IgG3 (Hu) Affinity Matrix can be used for the purification and isolation of IgG3, without cross-reactivity to other immunoglobulins.

CaptureSelect IgG3 (Hu) Affinity Matrix can be used for the purification and isolation of IgG3, without cross-reactivity to other immunoglobulins, like other human IgG subclasses, IgA, and IgM, from complex sources like plasma, serum, and cell culture supernatants.

Specifications

Content And Storage • Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light
Particle Size 35 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 5 mL
Type Affinity Matrix
Stationary Phase Anti-IgG3
Binding Capacity >6 g/L Resin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.