Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ KappaXP Affinity Matrix
GREENER_CHOICE

Catalog No. 2943212050 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
250 mL
1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-432-12050 50 mL
29-432-12005 5 mL
29-432-12010 10 mL
19-432-12250 250 mL
19-432-1201L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 29-432-12050 Supplier Thermo Scientific™ Supplier No. 2943212050
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Thermo Scientific™ CaptureSelect™ KappaXP Affinity Matrix is the improved version of CaptureSelect KappaXL Affinity Matrix, with a significant increase in binding capacity for both whole kappa monoclonal antibodies and kappa Fab's.

CaptureSelect KappaXP Affinity Matrix is the improved version of CaptureSelect KappaXL Affinity Matrix, with a significant increase in binding capacity for both whole kappa monoclonal antibodies and kappa Fab's. It enables efficient elution, even at pH 6.0, and demonstrates an improved stability for caustic cleaning agents. It is specific for human antibodies or antibody fragments, without cross-specificity to bovine antibody.

CaptureSelect KappaXP matrix recognizes the kappa light chain of human antibodies, enabling purification of all kappa-containing Ig's, Fab, and Fab2 fragments directly from complex sources and feed streams.

CaptureSelect KappaXP Affinity Matrix features include:

  • Purification of kappa monoclonal antibodies, Fab, and Fab2 fragments directly from complex source materials in a single step with high purity and yield
  • Mild elution for Fab fragments and antibodies
  • Human specific, no binding to bovine antibodies
  • Excellent scalability
  • Excellent pH stability up to 100 mM NaOH for limited exposure
  • Non–animal-derived

The high selectivity and high yields obtained using CaptureSelect KappaXP Affinity Matrix enable a robust and efficient purification process with excellent purity in one step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based
  • Average particle size: 65 μm
  • Ligand: CaptureSelect KappaXP affinity ligand
  • Ligand coupling method: epoxide immobilization of the ligand
  • Fab binding capacity: 20–30 g IgG Kappa-Fab/liter resin depending on flow rate, column height, and residence time
  • IgG binding capacity: 35–45 g IgG/liter resin depending on flow rate, column height, and residence time
  • Elution conditions: 20 mM citric acid or acetic acid, pH 3–4; 100 mM Tris, 1.5 M MgCl2, pH 6
  • Flow characteristics: 150–300 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Capacity (Metric) 35 g IgG/L matrix
Flow Rate 150 to 300 cm/hr. (Up to 2 bar)
Matrix Agarose-based
Content And Storage • Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light
Shipping Condition Wet Ice
Particle Size 65 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 50 mL
Type Affinity Matrix
Ligand Type KappaXP affinity ligand
Stationary Phase KappaXP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.