Learn More
Thermo Scientific™ CaptureSelect™ Lambda Fab Select Leakage ELISA
Shop All Thermo Scientific Products

Description
CaptureSelect LambdaFabSelect Leakage ELISA is designed to detect possible leached ligand of human Fab fragments or Ig's with a lambda light chain that have been purified using LambdaFabSelect affinity resin (Cytiva). Even when covalently attached, small amounts of the LambdaFabSelect affinity ligand can leach off of the chromatography support and co-elute with the antibody.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect LambdaFabSelect Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| Assay | ELISA |
| Detection Method | Absorbance at 450 nm |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Line | CaptureSelect |
| Product Type | Ligand Leakage ELISA |
| Sample Type | Ligand Leachate |
| Target Species | Not Applicable |
| For Use With (Equipment) | Microplate Reader |
| Quantity | 10 Assays |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.