Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ LC-kappa (mur) Affinity Matrix
GREENER_CHOICE

Catalog No. 191315050 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-131-5050 50 mL
19-131-5005 5 mL
19-131-5010 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-131-5050 Supplier Thermo Scientific™ Supplier No. 191315050
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Thermo Scientific™ CaptureSelect™ LC-kappa (mur) Affinity Matrix enables purification of murine (mouse, rat, and guinea pig) immunoglobulins and Fab fragments without cross reactivity towards bovine immunoglobulins.

CaptureSelect LC-kappa (mur) Affinity Matrix enables purification of murine (mouse, rat, and guinea pig) immunoglobulins and Fab fragments without cross reactivity towards bovine immunoglobulins. The ligand is directed towards a unique domain on the constant part of the kappa light chain of immunoglobulins, which makes CaptureSelect LC-kappa (mur) Affinity Matrix a unique resin in terms of specificity.

This matrix can be used for the purification and isolation of kappa light chain-containing murine immunoglobulins and Fab fragments from complex sources like plasma, serum, and cell culture supernatants in a single step. Due to its non-reactivity with bovine Ig, FCS can be applied in growth media without disturbing purification.

Specifications

Capacity (Metric) >15 g/L resin
Content And Storage • Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light
Particle Size 35 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 50 mL
Type Affinity Matrix
Stationary Phase Anti-LC-kappa (mur)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.