Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ MultiSpecies Albumin Depletion Product
GREENER_CHOICE

Catalog No. 191085350 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-108-5350 50 mL
19-108-5305 5 mL
19-108-5310 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-108-5350 Supplier Thermo Scientific™ Supplier No. 191085350
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This is one of a group of Thermo Scientific™ CaptureSelect™ Proteomics Toolbox™ affinity resins which are unique affinity resins developed for the depletion of proteins from human plasma or serum samples.

This is one of a group of CaptureSelect Proteomics Toolbox™ affinity resins which are unique affinity resins developed for the depletion of proteins from human plasma or serum samples. These affinity ligands are immobilized on a high capacity agarose support for easy, lower-pressure protein depletion or sample prep applications.

Specifications

Content And Storage • Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light
Particle Size 35 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 50 mL
Type Albumin Depletion Product
Stationary Phase Anti-albumin
Binding Capacity 15 to 25 mg Albumin/mL Resin
Can I use the High Select Top14 Abundant Protein Depletion Midi Spin Columns to deplete proteins from rodent samples?

The High Select Top14 Abundant Protein Depletion Midi Spin Columns have only been validated for human samples. We do not recommend using them for protein depletion in rodent samples. For protein depletion in rodent samples, we recommend the following multi-species depletion resins:

  • CaptureSelect MultiSpecies Albumin Depletion Product (Cat. No. 191085305)
  • Pierce Albumin Serum Depletion Kits (Cat. No. 89875)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.