Learn More
Thermo Scientific™ CaptureSelect™ Transferrin Affinity Matrix

Description
This is one of a group of CaptureSelect protein purification affinity resins developed for the simple, single-step purification of a variety human plasma and serum proteins. This unique selection of affinity purification resins can purify proteins or other non-antibody biomolecules from either recombinant or native sources with a high degree of affinity and specificity, and are immobilized on a high capacity agarose support.
Specifications
Specifications
| Content And Storage | Store at 2°C to 8°C. |
| Stationary Phase | Anti-transferrin |
| Product Line | CaptureSelect |
| Type | Affinity Matrix |
| Quantity | 50 mL |
| Form | Liquid Suspension |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.