Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ VII Select Leakage ELISA
GREENER_CHOICE

Catalog No. 810299001
Change view
Click to view available options
Quantity:
1 Assay
10 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81-029-9001 1 Assay
81-029-9010 10 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 81-029-9001 Supplier Thermo Scientific™ Supplier No. 810299001
Only null left
Add to Cart
Add to Cart
Request a Quote

Thermo Scientific™ CaptureSelect™ VIISelect Leakage ELISA is designed to detect possible leached ligand of human Factor VII purified using VIISelect affinity resin (Cytiva).

CaptureSelect VIISelect Leakage ELISA is designed to detect possible leached ligand of human Factor VII purified using VIISelect affinity resin (Cytiva). Even when covalently attached, small amounts of the VIISelect affinity ligand can leach off of the chromatography support and co-elute with the purified product.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect VIISelect Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

Specifications

Content And Storage Store at -5°C to -30°C.
Assay ELISA
For Use With (Equipment) Microplate Reader
Product Line CaptureSelect
Quantity 1 Assay
Target VIISelect Affinity Ligand
Type ELISA Kit
Format Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.