Learn More
Thomson Instrument Company Optimum Growth Vent Cap, Sterile
SDP
Catalog No. 501120010
Encompass_Preferred
Designed for use with Optimum Growth 1.6L, 2.8L and 5L Flasks, this sterile vent cap features a 0.2 µm PTFE membrane for enhanced aeration while maintaining sterility. Ideal for cell culture and microbial applications requiring gas exchange, it offers reliable performance and secure flask compatibility.Flask Compatibility: 1.6L, 2.8L, 5LMembrane: PTFEPore Size: 0.2µmSterility (SAL): 10⁻⁶ Made in the USA
Catalog No. 50-112-0010 Supplier Thomson Instrument Company Supplier No. 899116
Designed for use with Optimum Growth 1.6L, 2.8L and 5L Flasks, this sterile vent cap features a 0.2 µm PTFE membrane for enhanced aeration while maintaining sterility. Ideal for cell culture and microbial applications requiring gas exchange, it offers reliable performance and secure flask compatibility.Flask Compatibility: 1.6L, 2.8L, 5LMembrane: PTFEPore Size: 0.2µmSterility (SAL): 10⁻⁶ Made in the USA