Learn More
Aviva Systems Biology Corp CAPZA1 Antibody - N-terminal region (ARP64942_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64942_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CAPZA1. It was validated on Western Blot. . Alternate names: CAPPA1, CAPZ, CAZ1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-288-397