Learn More
Aviva Systems Biology Corp CAPZA2 Antibody - N-terminal region (ARP62108_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62108_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CAPZA2. It was validated on Western Blot. . Alternate names: CAPPA2, CAPZApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-287-570