Learn More
Aviva Systems Biology Corp Capzb Antibody - C-terminal region (ARP58436_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58436_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Capzb. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Rat Capzb. Alternate names: MGC95097ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-476